missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-piperidino-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 180403-13-8
Molecular Formula C9H12N2O2S
Molecular Weight (g/mol) 212.267
MDL Number MFCD11506352
InChI Key JIYHGZWZWANYAV-UHFFFAOYSA-N
Synonym 2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl
PubChem CID 33589513
IUPAC Name 2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid
SMILES C1CCN(CC1)C2=NC=C(S2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC64101CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC64101CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64101DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC64101DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

180403-13-8
212.267
JIYHGZWZWANYAV-UHFFFAOYSA-N
33589513
C1CCN(CC1)C2=NC=C(S2)C(=O)O
C9H12N2O2S
MFCD11506352
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid

Specifications

180403-13-8
100.0
Amber Glass Bottle
MFCD11506352
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl
C1CCN(CC1)C2=NC=C(S2)C(=O)O
212.267
212.27
2-Piperidino-1, 3-thiazole-5-carboxylic acid
96.5
97%
C9H12N2O2S
250mg
JIYHGZWZWANYAV-UHFFFAOYSA-N
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid
33589513
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'