missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Piperidineethanol, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC157610050

 View more versions of this product

Catalog No. AC157610050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Piperidineethanol
102°C
Glass bottle
234.0°C
38.0°C to 43.0°C
1484-84-0
C7H15NO
2-2-hydroxyethyl piperidine
PTHDBHDZSMGHKF-UHFFFAOYSA-N
2-piperidin-2-ylethanol
15144
Crystalline Low Melting Solid
1.0100g/mL
Authentic
1.01
White to Yellow
5g
94% min. (GC)
21,2
Solubility in water: very soluble. Other solubilities: soluble in alcohol
C1CCNC(C1)CCO
129.2
129.2
95%

Chemical Identifiers

1484-84-0
129.2
2-2-hydroxyethyl piperidine
2-piperidin-2-ylethanol
C7H15NO
PTHDBHDZSMGHKF-UHFFFAOYSA-N
15144
C1CCNC(C1)CCO
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 216-059-2

SDS Documents