missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Piperidineethanol, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157611000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Piperidineethanol | |
38.0°C to 43.0°C | |
1.0100g/mL | |
102°C | |
94% min. (GC) | |
C7H15NO | |
21,2 | |
2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol | |
PTHDBHDZSMGHKF-UHFFFAOYSA-N | |
2-piperidin-2-ylethanol | |
15144 | |
95% |
1484-84-0 | |
White to Yellow | |
234.0°C | |
Authentic | |
Glass bottle | |
100 g | |
1.01 | |
Solubility in water: very soluble. Other solubilities: soluble in alcohol | |
C1CCNC(C1)CCO | |
129.2 | |
129.2 | |
Crystalline Low Melting Solid |
Chemical Identifiers
1484-84-0 | |
129.2 | |
2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol | |
2-piperidin-2-ylethanol |
C7H15NO | |
PTHDBHDZSMGHKF-UHFFFAOYSA-N | |
15144 | |
C1CCNC(C1)CCO |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 216-059-2
RUO – Research Use Only