Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Piperazinone, 97%

$110.14 - $384.23

Chemical Identifiers

Chemical Name or Material 2-Piperazinone
CAS 5625-67-2
Molecular Formula C4H8N2O
Formula Weight 100.12
MDL Number MFCD01318687
Synonym 2-Oxopiperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2413103

  SDS
alfa aesar™
B2413103
1g Each for $110.14

AAB2413106

  SDS
alfa aesar
B2413106
5g Each for $384.23
Description & Specifications

Specifications

Formula Weight 100.12
Chemical Name or Material 2-Piperazinone
Melting Point 129° to 134°C
Health Hazard 1 Warning
Synonym 2-Oxopiperazine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 97%
Molecular Formula C4H8N2O
Health Hazard 2 GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
MDL Number MFCD01318687
CAS 5625-67-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.