missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piperazine-2-carboxylic acid dihydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 133660100

 View more versions of this product

Catalog No. AC133660100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Piperazine-2-carboxylic acid dihydrochloride
Glass bottle
200.0°C
3022-15-9
100.0
C5H10N2O2·2ClH
+/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, piperazin-2-carboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
C1CNC(CN1)C(=O)O.Cl.Cl
203.07
203.07
98%
Authentic
White to Yellow
10g
97.5
97.5 to 102.5% (Argentometry)
MFCD00044803
WNSDZBQLMGKPQS-UHFFFAOYSA-N
piperazine-2-carboxylic acid;dihydrochloride
2723757
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

3022-15-9
203.07
WNSDZBQLMGKPQS-UHFFFAOYSA-N
2723757
C1CNC(CN1)C(=O)O.Cl.Cl
C5H10N2O2·2ClH
MFCD00044803
+/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, piperazin-2-carboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
piperazine-2-carboxylic acid;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents