missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Picolyl chloride hydrochloride, 98%, ACROS Organics™

Manufacturer:  Acros Organics 172121000

 View more versions of this product

Catalog No. AC172121000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Picolyl chloride hydrochloride
119.0°C to 125.0°C
98%
97.5
Authentic
98%
C6H7Cl2N
2-(Chloromethyl)pyridine hydrochloride, 2-(Chloromethyl)pyridine hydrochloride
C1=CC=NC(=C1)CCl.Cl
164.029
CHEBI:76600
Crystalline Mass
Brown to Green-Yellow
100g
6959-47-3
100.0
Glass bottle
98%
MFCD00012811
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-(chloromethyl)pyridine;hydrochloride
23392
164.04

Chemical Identifiers

6959-47-3
164.029
JPMRGPPMXHGKRO-UHFFFAOYSA-N
23392
C6H7Cl2N
MFCD00012811
2-(Chloromethyl)pyridine hydrochloride, 2-(Chloromethyl)pyridine hydrochloride
CHEBI:76600
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Causes severe skin burns and eye damage.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

EINECSNumber : 230-149-9

RTECSNumber : US6825000

TSCA : TSCA

SDS Documents