Learn More
2-Picoline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A10847AP
Description
2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, sulfuric acid, acids, metals, iron(II) sulfate and hydrogen peroxide.
Specifications
2-Picoline | |
-70°C | |
128°C to 129°C | |
C6H7N | |
MFCD00006332 | |
UN2313 | |
14,7400 | |
Miscible with water. | |
CC1=CC=CC=N1 | |
93.129 | |
CHEBI:50415 | |
≥98% |
109-06-8 | |
0.944 | |
26°C (78°F) | |
1.5 | |
500 mL | |
104581 | |
2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha | |
BSKHPKMHTQYZBB-UHFFFAOYSA-N | |
2-methylpyridine | |
7975 | |
93.13 |
Chemical Identifiers
109-06-8 | |
93.129 | |
BSKHPKMHTQYZBB-UHFFFAOYSA-N | |
7975 | |
2-methylpyridine |
C6H7N | |
MFCD00006332 | |
2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha | |
CHEBI:50415 | |
CC1=CC=CC=N1 |
Safety and Handling
GHS H Statement
H226-H302-H312-H332-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
H226-H302+H332-H311-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES
EINECSNumber : 203-643-7
RTECSNumber : TJ4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only