Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phthalimidopropionic acid, 98%

$32.64 - $93.43

Chemical Identifiers

Chemical Name or Material 2-Phthalimidopropionic acid
CAS 21860-84-4
Molecular Formula C11H9NO4
Formula Weight 219.2
MDL Number MFCD00023061
Beilstein 86452
Synonym N-Phthaloyl-DL-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1353403

 
alfa aesar™
L1353403
1g Each for $32.64

AAL1353406

 
alfa aesar™
L1353406
5g Each for $93.43
Description & Specifications

Specifications

Formula Weight 219.2
Chemical Name or Material 2-Phthalimidopropionic acid
Assay 98%
Molecular Formula C11H9NO4
MDL Number MFCD00023061
Beilstein 86452
Synonym N-Phthaloyl-DL-alanine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
CAS 21860-84-4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 162° to 164°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.