Please login to your online account to display your discounted pricing

2-Phthalimidoethanesulfonyl chloride, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC430980050

 View more versions of this product

Catalog No. AC430980050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Slurry
Assay ≥94.0% (derivatized with diethylamine) (HPLC)
Formula Weight 273.7
Quantity 5g
Color Yellow to orange to brown
Melting Point 160° to 164°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 4403-36-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Chemical Name or Material 2-Phthalimidoethanesulfonyl chloride
Molecular Formula C10H8ClNO4S
MDL Number MFCD01861217
Packaging Glass bottle
Synonym 2-(1, 3-Dioxo-1, 3-dihydro-isoindol-2-yl)ethanesulfonyl chloride

Specifications

Chemical Name or Material 2-Phthalimidoethanesulfonyl chloride
CAS 4403-36-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0% (derivatized with diethylamine) (HPLC)
Assay Percent Range 95%
Molecular Formula C10H8ClNO4S
Formula Weight 273.7
MDL Number MFCD01861217
Synonym 2-(1, 3-Dioxo-1, 3-dihydro-isoindol-2-yl)ethanesulfonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents