Please login to your online account to display your discounted pricing

2-Phthalimidoethanesulfonyl chloride, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Phthalimidoethanesulfonyl chloride
CAS 4403-36-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0% (derivatized with diethylamine) (HPLC)
Assay Percent Range 95%
Molecular Formula C10H8ClNO4S
Formula Weight 273.7
MDL Number MFCD01861217
Synonym 2-(1, 3-Dioxo-1, 3-dihydro-isoindol-2-yl)ethanesulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430980010

  SDS
thermo fisher scientific
AC430980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94.0% (derivatized with diethylamine) (HPLC)
Formula Weight 273.7
Physical Form Slurry
Quantity 1g
Color Yellow to orange to brown
Melting Point 160° to 164°C
CAS Max % 100.0
CAS Min % 94.0
Molecular Formula C10H8ClNO4S
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Chemical Name or Material 2-Phthalimidoethanesulfonyl chloride
Assay Percent Range 95%
CAS 4403-36-5
MDL Number MFCD01861217
Packaging Glass bottle
Synonym 2-(1, 3-Dioxo-1, 3-dihydro-isoindol-2-yl)ethanesulfonyl chloride
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.