missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phosphonopropionic acid, 98+%

Manufacturer:  Alfa Aesar™ L1158804

 View more versions of this product

Catalog No. AAL1158804



Specifications

Specifications

2-Phosphonopropionic acid
118°C to 122°C
5962-41-4
MFCD00041415
1772834
GUXRZQZCNOHHDO-UHFFFAOYSA-N
2-phosphonopropanoic acid
73187
≥98%
Odorless
2g
C3H7O5P
UN3261
2-phosphonopropionic acid, acmc-20annb, phosphonopropionic acid, propanoic acid,2-phosphono, propanoic acid, 2-phosphono
CC(C(=O)O)P(=O)(O)O
154.058
154.06

Chemical Identifiers

5962-41-4
154.058
GUXRZQZCNOHHDO-UHFFFAOYSA-N
73187
CC(C(=O)O)P(=O)(O)O
C3H7O5P
MFCD00041415
2-phosphonopropionic acid, acmc-20annb, phosphonopropionic acid, propanoic acid,2-phosphono, propanoic acid, 2-phosphono
2-phosphonopropanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents