missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phosphonobutyric acid, 97%

Manufacturer:  Alfa Aesar™ L1203604

 View more versions of this product

Catalog No. AAL1203604



Specifications

Specifications

2-Phosphonobutyric acid
123°C to 127°C
4378-40-9
MFCD00041416
2-phosphonobutyric acid, phosphonobutyric acid, acmc-20annc, butanoic acid, 2-phosphono
CCC(C(=O)O)P(=O)(O)O
168.085
168.09
Odorless
2g
C4H9O5P
UN3261
PWSXRGRLZKVHLW-UHFFFAOYSA-N
2-phosphonobutanoic acid
4186724
97%

Chemical Identifiers

4378-40-9
168.085
PWSXRGRLZKVHLW-UHFFFAOYSA-N
4186724
CCC(C(=O)O)P(=O)(O)O
C4H9O5P
MFCD00041416
2-phosphonobutyric acid, phosphonobutyric acid, acmc-20annc, butanoic acid, 2-phosphono
2-phosphonobutanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents