Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phosphonobutyric acid, 97%


Quantity 10g

Manufacturer: alfa aesar™  L1203609

 View more versions of this product

Catalog No. AAL1203609

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.09
CAS 4378-40-9
Assay 97%
Chemical Name or Material 2-Phosphonobutyric acid
MDL Number MFCD00041416
Molecular Formula C4H9O5P
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Odorless
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN3261
Quantity 10g
Melting Point 123° to 127°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Specifications

Chemical Name or Material 2-Phosphonobutyric acid
CAS 4378-40-9
Molecular Formula C4H9O5P
Formula Weight 168.09
MDL Number MFCD00041416
UN Number UN3261
Accessories

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents