Please login to your online account to display your discounted pricing

2-Phenylthiophene, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Phenylthiophene
CAS 825-55-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C10H8S
Formula Weight 160.24
MDL Number MFCD00130080
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379590010

  SDS
thermo fisher scientific
AC379590010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379590050

  SDS
thermo fisher scientific
AC379590050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 160.24
Physical Form Crystalline Low Melting Mass
Quantity 1g
Color White to light beige
Melting Point 32° to 37°C
CAS Min % 95.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
MDL Number MFCD00130080
Assay Percent Range 96%
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 825-55-8
Molecular Formula C10H8S
Chemical Name or Material 2-Phenylthiophene
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.