missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylthiophene, 95%

Manufacturer:  Alfa Aesar™ H6081903

 View more versions of this product

Catalog No. AAH6081903



Specifications

Specifications

2-Phenylthiophene
256°C
1g
C10H8S
thiophene, 2-phenyl, 2-phenyl-thiophene, phenylthiophene, thiophene, phenyl, thiophene, 2-phenyl, 2-thienylbenzene, 2-phenyithiophene, 2-phenylthiofuran, 2-phenylthiophene, acmc-209ppe
C1=CC=C(C=C1)C2=CC=CS2
160.234
160.24
109°C (228°F)
34°C to 36°C
825-55-8
MFCD00130080
PJRGDKFLFAYRBV-UHFFFAOYSA-N
2-phenylthiophene
69999
95%

Chemical Identifiers

825-55-8
160.234
PJRGDKFLFAYRBV-UHFFFAOYSA-N
69999
C1=CC=C(C=C1)C2=CC=CS2
C10H8S
MFCD00130080
thiophene, 2-phenyl, 2-phenyl-thiophene, phenylthiophene, thiophene, phenyl, thiophene, 2-phenyl, 2-thienylbenzene, 2-phenyithiophene, 2-phenylthiofuran, 2-phenylthiophene, acmc-209ppe
2-phenylthiophene
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents