Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Phenylthio)ethanol, 99%

$33.24 - $101.45

Chemical Identifiers

Chemical Name or Material 2- (Phenylthio)ethanol
CAS 699-12-7
Molecular Formula C6H5S(CH2)2OH
Formula Weight 154.23
MDL Number MFCD00002905
Beilstein 1860057
Synonym 2-Hydroxyethyl phenyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1086109

  SDS
alfa aesar™
A1086109
10g Each for $33.24

AAA1086118

  SDS
alfa aesar™
A1086118
50g Each for $101.45
Description & Specifications

Specifications

Formula Weight 154.23
Chemical Name or Material 2- (Phenylthio)ethanol
TSCA TSCA
Assay 99%
Molecular Formula C6H5S(CH2)2OH
MDL Number MFCD00002905
CAS 699-12-7
Beilstein 1860057
Synonym 2-Hydroxyethyl phenyl sulfide
Flash Point >110°C (230°F)
Odor Unpleasant
Boiling Point 115° to 116°C (2mmHg)
Quantity 10g