Please login to your online account to display your discounted pricing

2-(Phenylthio)ethanimidamide hydrochloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Phenylthio)ethanimidamide hydrochloride
CAS 84544-86-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2S·ClH
Linear Formula C6H5SCH2C(=NH)NH2·HCl
Formula Weight 202.71
MDL Number MFCD00100031
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05877EA

  SDS
fisher scientific company
SPB05877EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB05877DA

  SDS
fisher scientific company
SPB05877DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.71
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-(Phenylthio)ethanimidamide hydrochloride
Packaging Amber glass bottle
Molecular Formula C8H10N2S·ClH
CAS 84544-86-5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Assay Percent Range 97%
Linear Formula C6H5SCH2C(=NH)NH2·HCl
MDL Number MFCD00100031
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.