Please login to your online account to display your discounted pricing

2-(Phenylthio)ethanimidamide hydrochloride, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SPB05877DA

 View more versions of this product

Catalog No. SPB05877DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.71
Quantity 1g
Assay 97%
CAS Min % 97.0
Linear Formula C6H5SCH2C(=NH)NH2·HCl
Molecular Formula C8H10N2S·ClH
CAS 84544-86-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
MDL Number MFCD00100031
Packaging Amber glass bottle
Chemical Name or Material 2-(Phenylthio)ethanimidamide hydrochloride
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-(Phenylthio)ethanimidamide hydrochloride
CAS 84544-86-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2S·ClH
Linear Formula C6H5SCH2C(=NH)NH2·HCl
Formula Weight 202.71
MDL Number MFCD00100031
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents