Please login to your online account to display your discounted pricing

2-(Phenylthio)ethanethioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Phenylthio)ethanethioamide
CAS 59865-82-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H9NS2
Linear Formula C6H5SCH2CSNH2
Formula Weight 183.3
MDL Number MFCD00085154
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04192EA

  SDS
fisher scientific company
KM04192EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.3
Quantity 10g
Assay 95%
CAS Min % 95.0
Linear Formula C6H5SCH2CSNH2
Packaging Amber glass bottle
Chemical Name or Material 2-(Phenylthio)ethanethioamide
Assay Percent Range 95%
Molecular Formula C8H9NS2
MDL Number MFCD00085154
CAS 59865-82-6