Please login to your online account to display your discounted pricing

2-(Phenylthio)ethanethioamide, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM04192EA

 View more versions of this product

Catalog No. KM04192EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 183.3
Quantity 10g
Assay 95%
CAS Min % 95.0
Packaging Amber glass bottle
Linear Formula C6H5SCH2CSNH2
Assay Percent Range 95%
Chemical Name or Material 2-(Phenylthio)ethanethioamide
MDL Number MFCD00085154
Molecular Formula C8H9NS2
CAS 59865-82-6

Specifications

Chemical Name or Material 2-(Phenylthio)ethanethioamide
CAS 59865-82-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H9NS2
Linear Formula C6H5SCH2CSNH2
Formula Weight 183.3
MDL Number MFCD00085154