Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylthiazolidine, 97%

Chemical Identifiers

Chemical Name or Material 2-Phenylthiazolidine
CAS 4569-82-8
Formula Weight 165.26
MDL Number MFCD00128238
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6282306

 
alfa aesar™
H6282306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6282303

 
alfa aesar™
H6282303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62823MD

 
alfa aesar™
H62823MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.26
Chemical Name or Material 2-Phenylthiazolidine
Physical Form Solid
Assay 97%
MDL Number MFCD00128238
CAS 4569-82-8
RTECS Number XJ6094000
Quantity 5g