missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylthiazole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H54521MD

 View more versions of this product

Catalog No. AAH54521MD



Specifications

Specifications

2-Phenylthiazole-4-carboxylic acid
250mg
C10H6NO2S
2-phenyl-1,3-thiadiazole-4-carboxylic acid
C1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
204.22
205.23
148°C to 152°C
7113-10-2
MFCD00141954
IBUSLNJQKLZPNR-UHFFFAOYSA-M
2-phenyl-1,3-thiazole-4-carboxylic acid
138928
97%

Chemical Identifiers

7113-10-2
204.22
IBUSLNJQKLZPNR-UHFFFAOYSA-M
138928
C1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
C10H6NO2S
MFCD00141954
2-phenyl-1,3-thiadiazole-4-carboxylic acid
2-phenyl-1,3-thiazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents