missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylthiazole-4-acetic acid, 97%

$42.58 - $556.65

Chemical Identifiers

CAS 16441-28-4
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD01312954
InChI Key LYHDWKGJPJRCTG-UHFFFAOYSA-N
Synonym 2-2-phenyl-1,3-thiazol-4-yl acetic acid
PubChem CID 701757
IUPAC Name 2-(2-phenyl-1,3-thiazol-4-yl)acetic acid
SMILES C1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54805MD View Documents Alfa Aesar™
H54805MD
250mg Each for $42.58
 
AAH5480503 View Documents Alfa Aesar™
H5480503
1g Each for $134.36
 
AAH5480506 View Documents Alfa Aesar™
H5480506
5g Each for $556.65
 
Specifications

Chemical Identifiers

16441-28-4
219.258
LYHDWKGJPJRCTG-UHFFFAOYSA-N
701757
C1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O
C11H9NO2S
MFCD01312954
2-2-phenyl-1,3-thiazol-4-yl acetic acid
2-(2-phenyl-1,3-thiazol-4-yl)acetic acid

Specifications

250mg
C11H9NO2S
2-2-phenyl-1,3-thiazol-4-yl acetic acid
C1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O
219.258
219.26
2-Phenylthiazole-4-acetic acid
16441-28-4
MFCD01312954
LYHDWKGJPJRCTG-UHFFFAOYSA-N
2-(2-phenyl-1,3-thiazol-4-yl)acetic acid
701757
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures