missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylthiazole-4-acetic acid, 97%

Manufacturer:  Alfa Aesar™ H5480503

 View more versions of this product

Catalog No. AAH5480503



Specifications

Specifications

2-Phenylthiazole-4-acetic acid
16441-28-4
MFCD01312954
LYHDWKGJPJRCTG-UHFFFAOYSA-N
2-(2-phenyl-1,3-thiazol-4-yl)acetic acid
701757
97%
1g
C11H9NO2S
2-2-phenyl-1,3-thiazol-4-yl acetic acid
C1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O
219.258
219.26

Chemical Identifiers

16441-28-4
219.258
LYHDWKGJPJRCTG-UHFFFAOYSA-N
701757
C1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O
C11H9NO2S
MFCD01312954
2-2-phenyl-1,3-thiazol-4-yl acetic acid
2-(2-phenyl-1,3-thiazol-4-yl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents