Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Phenylsulfonyl)thiophene, 98%

$96.97 - $397.14

Chemical Identifiers

Chemical Name or Material 2- (Phenylsulfonyl)thiophene
CAS 22407-40-5
Molecular Formula C10H8O2S2
Formula Weight 224.3
MDL Number MFCD00052142
Beilstein 146279
Synonym 2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1076903

 
alfa aesar™
A1076903
1g Each for $96.97

AAA1076906

 
alfa aesar™
A1076906
5g Each for $397.14
Description & Specifications

Specifications

Formula Weight 224.3
Chemical Name or Material 2- (Phenylsulfonyl)thiophene
Assay 98%
Molecular Formula C10H8O2S2
MDL Number MFCD00052142
CAS 22407-40-5
Beilstein 146279
Synonym 2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone
Melting Point 121° to 125°C
Quantity 1g