Please login to your online account to display your discounted pricing

2-(Phenylsulfonyl)aniline, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Phenylsulfonyl)aniline
CAS 4273-98-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12 H11 N O2 S
Formula Weight 233.28
MDL Number MFCD00007706
Synonym 2-Aminophenyl phenyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209020050

  SDS
thermo fisher scientific
AC209020050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.28
Physical Form Powder
Assay ≥98.5%
Quantity 5g
Color Off-white to beige
Melting Point 118° to 122°C
Beilstein 13, 399
CAS Max % 100.0
CAS Min % 98.5
Assay Percent Range 99%
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 2-Aminophenyl phenyl sulfone
Molecular Formula C12 H11 N O2 S
Chemical Name or Material 2-(Phenylsulfonyl)aniline
CAS 4273-98-7
MDL Number MFCD00007706
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.