Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Phenylsulfonyl)acetophenone, 99%

$140.74 - $140.74

Chemical Identifiers

Chemical Name or Material 2-(Phenylsulfonyl)acetophenone
CAS 3406-03-9
Molecular Formula C14H12O3S
Formula Weight 260.31
MDL Number MFCD00025043
Beilstein 1879574
Synonym 2-(Benzenesulfonyl)acetophenone; Phenacyl phenyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0503214

  SDS
alfa aesar™
L0503214
25g Each for $140.74
Description & Specifications

Specifications

Formula Weight 260.31
Chemical Name or Material 2-(Phenylsulfonyl)acetophenone
Molecular Formula C14H12O3S
MDL Number MFCD00025043
CAS 3406-03-9
Synonym 2-(Benzenesulfonyl)acetophenone; Phenacyl phenyl sulfone
Assay 99%
Beilstein 1879574
Melting Point 92° to 96°C
Quantity 25g