Please login to your online account to display your discounted pricing

2-Phenylquinoline, +99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  297990010

 View more versions of this product

Catalog No. AC297990010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 205.26
Physical Form Crystalline Powder
Quantity 1g
Color White to light yellow
Melting Point 82° to 84°C
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Assay Percent Range 99+%
CAS 612-96-4
Molecular Formula C15H11N
Chemical Name or Material 2-Phenylquinoline
MDL Number MFCD00011568
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Phenylquinoline
CAS 612-96-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C15H11N
Formula Weight 205.26
MDL Number MFCD00011568