missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylpropylamine hydrochloride, ≥98%

Manufacturer:  Alfa Aesar™ H3633822

 View more versions of this product

Catalog No. AAH3633822


 

Specifications

Specifications

2-Phenylpropylamine hydrochloride
Cream to Yellow
100g
≥98%
MFCD01708026
HBVYOCJBEXSCQE-UHFFFAOYSA-N
2-phenylpropan-1-amine;hydrochloride
89332
Powder
Hygroscopic
143°C to 145°C
20388-87-8
C9H14ClN
β-Methylphenylamine hydrochloride
CC(CN)C1=CC=CC=C1.Cl
171.668
171.67

Chemical Identifiers

20388-87-8
171.668
HBVYOCJBEXSCQE-UHFFFAOYSA-N
89332
C9H14ClN
MFCD01708026
β-Methylphenylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents