Please login to your online account to display your discounted pricing

2-Phenylpropyl isobutyrate, 98%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  183850250

 View more versions of this product

Catalog No. AC183850250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 206.28
Physical Form Liquid
Quantity 25g
Color Clear colorless
TSCA TSCA
EINECS Number 265-931-9
CAS Max % 100.0
CAS Min % 97.5
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula (CH3)2CHCO2CH2CH(C6H5)CH3
MDL Number MFCD00008915
Name Note 98%
Chemical Name or Material 2-Phenylpropyl isobutyrate
CAS 65813-53-8
Assay Percent Range 98%
Molecular Formula C13H18O2

Specifications

Chemical Name or Material 2-Phenylpropyl isobutyrate
Name Note 98%
CAS 65813-53-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H18O2
Linear Formula (CH3)2CHCO2CH2CH(C6H5)CH3
Formula Weight 206.28
MDL Number MFCD00008915