Please login to your online account to display your discounted pricing

2-Phenylpropyl isobutyrate, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Phenylpropyl isobutyrate
Name Note 98%
CAS 65813-53-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H18O2
Linear Formula (CH3)2CHCO2CH2CH(C6H5)CH3
Formula Weight 206.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC183850250

  SDS
thermo fisher scientific
183850250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC183850050

  SDS
thermo fisher scientific
183850050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 206.28
Quantity 25g
Color Clear colorless
EINECS Number 265-931-9
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2-Phenylpropyl isobutyrate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C13H18O2
Linear Formula (CH3)2CHCO2CH2CH(C6H5)CH3
MDL Number MFCD00008915
CAS 65813-53-8
Infrared Spectrum Authentic
CAS Min % 97.5
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)