Please login to your online account to display your discounted pricing

2-Phenylpiperidine, 97%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC450850010

 View more versions of this product

Catalog No. AC450850010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.25
Quantity 1g
Chemical Name or Material 2-Phenylpiperidine
Assay Percent Range 97%
Molecular Formula C11H15N
CAS 3466-80-6

Specifications

Chemical Name or Material 2-Phenylpiperidine
Assay Percent Range 97%
Molecular Formula C11H15N
Formula Weight 161.25