Please login to your online account to display your discounted pricing

2-Phenylpiperidine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Phenylpiperidine
Assay Percent Range 97%
Molecular Formula C11H15N
Formula Weight 161.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC450850050

 
thermo fisher scientific
AC450850050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450850010

 
thermo fisher scientific
AC450850010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.25
Quantity 5g
Chemical Name or Material 2-Phenylpiperidine
Assay Percent Range 97%
Molecular Formula C11H15N
CAS 3466-80-6