missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylphenol, 99%

$49.20 - $189.68

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
IUPAC Name 2-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=CC=C2O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA105920E View Documents Alfa Aesar™
A105920E
2500g Each for $189.68
 
AAA1059230 View Documents Alfa Aesar™
A1059230
250g Each for $49.20
 
AAA1059236 View Documents Alfa Aesar™
A1059236
500g Each for $55.05
 
Description

Description

Applications
2-Phenylphenol is used as a dye intermediate, plasticizer, rubber chemical and as a preservative in water-oil emulsions. It is used as an antimicrobial preservative in cosmetics and an antimicrobial additive in the manufacture of metalworking fluids, leather, adhesives and textiles. Further, it serves as a disinfectant in households, hospitals, nursing homes, farms, laundries, barber shops, veterinary equipment and food processing plants. It is also employed in the production of resins and rubber chemicals. In addition to this, it is used as a polymer material, stabilizer and flame retardant.

Solubility
Soluble in ethanol, acetone, benzene,sodium hydroxide, chloroform, acetonitrile, toluene, hexane, ligroin, ethyl ether, pyridine, ethylene glycol, isopropanol, glycol ethers and polyglycols.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

90-43-7
170.211
LLEMOWNGBBNAJR-UHFFFAOYSA-N
7017
2-phenylphenol
C12H10O
MFCD00002208
2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
CHEBI:17043
C1=CC=C(C=C1)C2=CC=CC=C2O

Specifications

124°C (255°F)
152°C to 154°C (15mmHg)
2500g
C12H10O
UN3077
14,7304
LLEMOWNGBBNAJR-UHFFFAOYSA-N
2-phenylphenol
7017
170.21
2-Phenylphenol
Mild
54°C to 58°C
90-43-7
MFCD00002208
606907
2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
C1=CC=C(C=C1)C2=CC=CC=C2O
170.211
CHEBI:17043
99%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : DV5775000

TSCA : TSCA