Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylindole-3-carboxaldehyde, 97%

$47.41 - $644.96

Chemical Identifiers

Chemical Name or Material 2-Phenylindole-3-carboxaldehyde
CAS 25365-71-3
Molecular Formula C15H11NO
Formula Weight 221.26
MDL Number MFCD00435481
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2104103

 
alfa aesar™
B2104103
1g Each for $47.41

AAB2104106

 
alfa aesar™
B2104106
5g Each for $163.16

AAB2104114

 
alfa aesar™
B2104114
25g Each for $644.96
Description & Specifications

Specifications

Formula Weight 221.26
Chemical Name or Material 2-Phenylindole-3-carboxaldehyde
Assay 97%
Molecular Formula C15H11NO
MDL Number MFCD00435481
CAS 25365-71-3
Melting Point 249° to 253°C
Quantity 1g