Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Phenyliminomethyl)thiophene-5-boronic acid pinacol ester, 95%

$201.64 - $201.64

Chemical Identifiers

Chemical Name or Material 2-(Phenyliminomethyl)thiophene-5-boronic acid pinacol ester
CAS 1257651-22-1
Molecular Formula C17H20BNO2S
Formula Weight 313.22g/mol
MDL Number MFCD16294526
Synonym Phenyl[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienylmethylene]amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5189906

 
alfa aesar™
H51899-06
Each for $201.64
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 2-(Phenyliminomethyl)thiophene-5-boronic acid pinacol ester
CAS 1257651-22-1
Assay 95%
Molecular Formula C17H20BNO2S
Formula Weight 313.22g/mol
MDL Number MFCD16294526
Synonym Phenyl[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienylmethylene]amine