missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethylamine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441911000

 View more versions of this product

Catalog No. AC441911000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Phenylethylamine hydrochloride
Consistent with structure
220°C to 223°C
156-28-5
100.0
C8H11N·ClH
1-phenyl-2-aminoethane hydrochloride, 2-phenylethanamine hydrochloride, 2-phenylethylamine hydrochloride, benzeneethanamine, hydrochloride, beta-phenethylamine hydrochloride, beta-phenylethylamine hydrochloride, phenethylamine hydrochloride, phenethylammonium chloride, unii-vof61xw69d, usaf el-76
C1=CC=C(C=C1)CCN.Cl
157.64
157.64
98%
Authentic
White
100g
97.5
97.5 to max. 102.5% min. (Argentometry)
15,7339
SKHIBNDAFWIOPB-UHFFFAOYSA-N
2-phenylethanamine;hydrochloride
9075
Powder or Crystalline Powder

Chemical Identifiers

156-28-5
157.64
1-phenyl-2-aminoethane hydrochloride, 2-phenylethanamine hydrochloride, 2-phenylethylamine hydrochloride, benzeneethanamine, hydrochloride, beta-phenethylamine hydrochloride, beta-phenylethylamine hydrochloride, phenethylamine hydrochloride, phenethylammonium chloride, unii-vof61xw69d, usaf el-76
2-phenylethanamine;hydrochloride
C8H11N·ClH
SKHIBNDAFWIOPB-UHFFFAOYSA-N
9075
C1=CC=C(C=C1)CCN.Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-849-2

SDS Documents