Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylethyl propionate, 97%

$42.34 - $147.81

Chemical Identifiers

Chemical Name or Material 2-Phenylethyl propionate
CAS 122-70-3
Molecular Formula C6H5(CH2)2OCOC2H5
Formula Weight 178.22
MDL Number MFCD00027008
Synonym Phenethyl propionate; Propionic acid 2-phenylethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2128318

  SDS
alfa aesar™
B2128318
50g Each for $42.34

AAB2128330

  SDS
alfa aesar™
B2128330
250g Each for $147.81
Description & Specifications

Specifications

Formula Weight 178.22
Chemical Name or Material 2-Phenylethyl propionate
TSCA TSCA
Odor Floral
Assay 97%
Molecular Formula C6H5(CH2)2OCOC2H5
MDL Number MFCD00027008
CAS 122-70-3
Synonym Phenethyl propionate; Propionic acid 2-phenylethyl ester
Boiling Point 245°C
Flash Point 106°C (223°F)
RTECS Number UF6488000
Quantity 50g