missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2-Phenylethyl)phosphonic Acid 98.0+%, TCI America™
$368.30 - $1284.06
Chemical Identifiers
CAS | 4672-30-4 |
---|---|
Molecular Formula | C8H11O3P |
Molecular Weight (g/mol) | 186.147 |
MDL Number | MFCD00485102 |
InChI Key | BFDYSJCMAFSRDH-UHFFFAOYSA-N |
Synonym | Phenethylphosphonic Acid |
PubChem CID | 78404 |
IUPAC Name | 2-phenylethylphosphonic acid |
SMILES | C1=CC=C(C=C1)CCP(=O)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P2122200MG
|
TCI America
P2122200MG |
200 mg |
Each for $368.30
|
|
|||||
P21221G
|
TCI America
P21221G |
1 g |
Each for $1,284.06
|
|
|||||
Chemical Identifiers
4672-30-4 | |
186.147 | |
BFDYSJCMAFSRDH-UHFFFAOYSA-N | |
78404 | |
C1=CC=C(C=C1)CCP(=O)(O)O |
C8H11O3P | |
MFCD00485102 | |
Phenethylphosphonic Acid | |
2-phenylethylphosphonic acid |
Specifications
4672-30-4 | |
C8H11O3P | |
200 mg | |
BFDYSJCMAFSRDH-UHFFFAOYSA-N | |
2-phenylethylphosphonic acid | |
78404 | |
≥98.0% (HPLC,T) | |
(2-Phenylethyl)phosphonic Acid |
138°C | |
MFCD00485102 | |
Phenethylphosphonic Acid | |
C1=CC=C(C=C1)CCP(=O)(O)O | |
186.147 | |
186.15 | |
Crystalline Powder |
Safety and Handling
TSCA : No