missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethyl Isovalerate, TCI America™

Manufacturer:  TCI America P2009500ML

 View more versions of this product

Catalog No. P2009500ML



Specifications

Specifications

2-Phenylethyl Isovalerate
Colorless
140-26-1
C13H18O2
Isovaleric Acid 2-Phenylethyl Ester, Phenethyl Isovalerate, Isovaleric Acid Phenethyl Ester
CC(C)CC(=O)OCCC1=CC=CC=C1
206.285
206.29
268°C
500mL
≥98.0% (GC)
MFCD00044322
JIMGVOCOYZFDKB-UHFFFAOYSA-N
2-phenylethyl 3-methylbutanoate
8792
Clear Liquid at 20°C

Chemical Identifiers

140-26-1
206.285
JIMGVOCOYZFDKB-UHFFFAOYSA-N
8792
CC(C)CC(=O)OCCC1=CC=CC=C1
C13H18O2
MFCD00044322
Isovaleric Acid 2-Phenylethyl Ester, Phenethyl Isovalerate, Isovaleric Acid Phenethyl Ester
2-phenylethyl 3-methylbutanoate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1040&(3)-1045

RTECSNumber : NY1511500

TSCA : Yes

Documents