missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylethyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A1512322

 View more versions of this product

Catalog No. AAA1512322



Description

Description

Applications
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.
Specifications

Specifications

2-Phenylethyl isothiocyanate
Pungent
100g
C9H9NS
UN2922
2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
C1=CC=C(C=C1)CCN=C=S
163.24
CHEBI:351346
98%
>110°C (230°F)
139°C to 140°C (11mmHg)
2257-09-2
MFCD00004821
2084162
IZJDOKYDEWTZSO-UHFFFAOYSA-N
2-isothiocyanatoethylbenzene
16741
163.24

Chemical Identifiers

2257-09-2
163.24
IZJDOKYDEWTZSO-UHFFFAOYSA-N
16741
2-isothiocyanatoethylbenzene
C9H9NS
MFCD00004821
2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
CHEBI:351346
C1=CC=C(C=C1)CCN=C=S
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : NX9115000

TSCA : TSCA

Documents