missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethanethiol, TCI America™

Manufacturer:  TCI America P171525G

 View more versions of this product

Catalog No. P171525G



Specifications

Specifications

2-Phenylethanethiol
Yellow
4410-99-5
MFCD00004891
ZMRFRBHYXOQLDK-UHFFFAOYSA-N
2-phenylethanethiol
78126
Clear Liquid at 20°C
218°C
25g
C8H10S
phenethyl mercaptan, benzeneethanethiol, 2-phenylethyl mercaptan, phenylethyl mercaptan, 2-phenylethylthiol, unii-i4th2t2iny, 2-phenyl-ethanethiol, 2-phenylethylmercaptan, beta-phenylethyl mercaptan, i4th2t2iny
C1=CC=C(C=C1)CCS
138.228
138.23
≥97.0% (GC,T)

Chemical Identifiers

4410-99-5
138.228
ZMRFRBHYXOQLDK-UHFFFAOYSA-N
78126
C1=CC=C(C=C1)CCS
C8H10S
MFCD00004891
phenethyl mercaptan, benzeneethanethiol, 2-phenylethyl mercaptan, phenylethyl mercaptan, 2-phenylethylthiol, unii-i4th2t2iny, 2-phenyl-ethanethiol, 2-phenylethylmercaptan, beta-phenylethyl mercaptan, i4th2t2iny
2-phenylethanethiol
Safety and Handling

Safety and Handling

TSCA : No

Documents