missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylbutyryl chloride, 98%

$123.24 - $432.04

Chemical Identifiers

CAS 36854-57-6
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00018811
InChI Key QGXMHCMPIAYMGT-UHFFFAOYSA-N
Synonym .alpha.-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenyl butyryl chloride, 2-phenyl-2-ethylacetyl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, 2-phenylbutyryl chloride, 2-phenylbutyrylchloride, acmc-209ipa, butyryl chloride, 2-phenyl
PubChem CID 98173
IUPAC Name 2-phenylbutanoyl chloride
SMILES CCC(C1=CC=CC=C1)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2299714
View Documents
Alfa Aesar™
B2299714
25g
Each for $123.24
 
AAB2299722
View Documents
Alfa Aesar™
B2299722
100g
Each for $432.04
 
Description

Description

Specifications

Chemical Identifiers

36854-57-6
182.647
QGXMHCMPIAYMGT-UHFFFAOYSA-N
98173
CCC(C1=CC=CC=C1)C(=O)Cl
C10H11ClO
MFCD00018811
.alpha.-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenyl butyryl chloride, 2-phenyl-2-ethylacetyl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, 2-phenylbutyryl chloride, 2-phenylbutyrylchloride, acmc-209ipa, butyryl chloride, 2-phenyl
2-phenylbutanoyl chloride

Specifications

1.093
1.517
143°C to 145°C (15mmHg)
36854-57-6
MFCD00018811
2045209
QGXMHCMPIAYMGT-UHFFFAOYSA-N
2-phenylbutanoyl chloride
98173
98%
98°C (208°F)
Moisture sensitive
25g
C10H11ClO
UN3265
.alpha.-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenyl butyryl chloride, 2-phenyl-2-ethylacetyl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, 2-phenylbutyryl chloride, 2-phenylbutyrylchloride, acmc-209ipa, butyryl chloride, 2-phenyl
CCC(C1=CC=CC=C1)C(=O)Cl
182.647
182.65
2-Phenylbutyryl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 253-241-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS