Please login to your online account to display your discounted pricing

2-Phenylbutyryl chloride, 96%, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346000250

 View more versions of this product

Catalog No. AC346000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 182.65
Quantity 25g
Color Clear pale yellow to yellow
Beilstein 09, IV, 1817
EINECS Number 253-241-0
TSCA TSCA
Density 1.093g/mL
CAS Max % 100.0
CAS Min % 95.0
Refractive Index 1.5160 to 1.5180 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 96%
Molecular Formula C10H11ClO
CAS 36854-57-6
Linear Formula C6H5CH(CHCH3)C(O)Cl
Chemical Name or Material 2-Phenylbutyryl chloride
MDL Number MFCD00018811

Specifications

Chemical Name or Material 2-Phenylbutyryl chloride
CAS 36854-57-6
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C10H11ClO
Linear Formula C6H5CH(CHCH3)C(O)Cl
Formula Weight 182.65
MDL Number MFCD00018811
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents