Please login to your online account to display your discounted pricing

2-Phenylbutyric Acid 98%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  130375000

 View more versions of this product

Catalog No. AC130375000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Adhering Crystals
Assay ≥97.5%
Formula Weight 164.2
Quantity 500g
Color White to yellow
EINECS Number 201-982-5
TSCA TSCA
RTECS Number ET5957500
Melting Point 39° to 42°C
Beilstein 09, II, 356
CAS Max % 100.0
Chemical Name or Material 2-Phenylbutyric acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H12O2
Linear Formula C2H5CH(C6H5)CO2H
MDL Number MFCD00002667
CAS 90-27-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Phenylbutyric acid
Name Note 98%
CAS 90-27-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O2
Linear Formula C2H5CH(C6H5)CO2H
Formula Weight 164.2
MDL Number MFCD00002667
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents