Please login to your online account to display your discounted pricing

2-Phenylbutyric Acid 98%, ACROS Organics™

$171.58 - $171.58

Chemical Identifiers

Chemical Name or Material 2-Phenylbutyric acid
Name Note 98%
CAS 90-27-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O2
Linear Formula C2H5CH(C6H5)CO2H
Formula Weight 164.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC130375000

  SDS
thermo fisher scientific
130375000
500g Each for $171.58
Description & Specifications

Specifications

Physical Form Adhering Crystals
Assay ≥97.5%
Formula Weight 164.2
Quantity 500g
Color White to yellow
EINECS Number 201-982-5
TSCA TSCA
RTECS Number ET5957500
Melting Point 39° to 42°C
Beilstein 09, II, 356
CAS Max % 100.0
Chemical Name or Material 2-Phenylbutyric acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H12O2
Linear Formula C2H5CH(C6H5)CO2H
MDL Number MFCD00002667
CAS 90-27-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.