Please login to your online account to display your discounted pricing

2-Phenylbutyramide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  348800050

 View more versions of this product

Catalog No. AC348800050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Needles
Formula Weight 163.22
Quantity 5g
Color White
EINECS Number 201-980-4
TSCA TSCA
RTECS Number ES4955000
Melting Point 83° to 87°C
Merck Index 13, 7362
CAS Max % 100.0
Chemical Name or Material 2-Phenylbutyramide
Assay Percent Range 98%
Molecular Formula C10H13NO
Linear Formula H2NCOCH(C2H5)C6H5
MDL Number MFCD00025511
CAS 90-26-6
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Phenylbutyramide
CAS 90-26-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H13NO
Linear Formula H2NCOCH(C2H5)C6H5
Formula Weight 163.22
MDL Number MFCD00025511