Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylbutyramide, 97%

$54.85 - $195.88

Chemical Identifiers

Chemical Name or Material 2-Phenylbutyramide
CAS 90-26-6
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD00025511
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1850122

  SDS
alfa aesar™
A1850122
100g Each for $54.85

AAA1850136

  SDS
alfa aesar™
A1850136
500g Each for $195.88
Description & Specifications

Specifications

Formula Weight 163.22
Chemical Name or Material 2-Phenylbutyramide
Assay 97%
Molecular Formula C10H13NO
MDL Number MFCD00025511
CAS 90-26-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ES4955000
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.