Please login to your online account to display your discounted pricing

2-Phenylbenzoxazole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Phenylbenzoxazole
CAS 833-50-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13 H9 N O
Formula Weight 195.22
MDL Number MFCD00012183
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305150010

  SDS
thermo fisher scientific
305150010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 195.22
Quantity 1g
Color Light yellow-beige
Melting Point 100° to 103°C
CAS Max % 100.0
Chemical Name or Material 2-Phenylbenzoxazole
Molecular Formula C13 H9 N O
CAS 833-50-1
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
MDL Number MFCD00012183
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.