missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Phenylbenzimidazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1099606
Description
2-Phenylbenzimidazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Phenylbenzimidazole is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Phenylbenzimidazole | |
296°C to 297°C | |
MFCD00005592 | |
7087 | |
2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole | |
DWYHDSLIWMUSOO-UHFFFAOYSA-N | |
2-phenyl-1H-1,3-benzodiazole | |
12855 | |
97% |
716-79-0 | |
C13H10N2 | |
5 g | |
14,7274 | |
Slightly soluble in water. | |
N1C2=CC=CC=C2N=C1C1=CC=CC=C1 | |
194.24 | |
194.24 |
Chemical Identifiers
716-79-0 | |
194.24 | |
DWYHDSLIWMUSOO-UHFFFAOYSA-N | |
12855 |
C13H10N2 | |
MFCD00005592 | |
2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole | |
N1C2=CC=CC=C2N=C1C1=CC=CC=C1 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 211-939-2
RTECSNumber : DE0360000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only